Assay Details
| Gene Symbol: RPUSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075874.1 | NP_001069342.1 | 5 - 6 | 666 | 70 | |
| GenBank mRNA | BC118196.1 | - | 5 - 6 | 666 | 70 | |
More Information
Set Membership: |
|

RPUSD1
Genomic Map