Assay Details
| Gene Symbol: TAF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075975.1 | NP_001069443.1 | 1 - 2 | 242 | 151 | |
| GenBank mRNA | BC118236.1 | - | 1 - 2 | 242 | 151 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118236.1, NM_001075975.1 transcripts.
Set Membership: |
|

TAF9
Genomic Map