Assay Details
| Gene Symbol: SPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076003.1 | NP_001069471.1 | 2 - 3 | 633 | 76 | |
| GenBank mRNA | BC118299.1 | - | 2 - 3 | 633 | 76 | |
| DQ978331.1 | - | 2 - 3 | 642 | 76 | ||
More Information
Set Membership: |
|

SPR
Genomic Map