Assay Details
| Gene Symbol: DUSP26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076004.1 | NP_001069472.1 | 2 - 3 | 500 | 56 | |
| GenBank mRNA | BC118329.1 | - | 2 - 3 | 500 | 56 | |
More Information
Set Membership: |
|

DUSP26
Genomic Map