Assay Details
| Gene Symbol: TFDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076029.1 | NP_001069497.1 | 10 - 11 | 1124 | 65 | |
| GenBank mRNA | BC118103.1 | - | 10 - 11 | 1124 | 65 | |
More Information
Set Membership: |
|

TFDP1
Genomic Map