Assay Details
| Gene Symbol: SNRNP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076046.2 | NP_001069514.1 | 4 - 5 | 436 | 65 | |
| GenBank mRNA | BC114762.1 | - | 4 - 5 | 437 | 65 | |
More Information
Set Membership: |
|

SNRNP70
Genomic Map