Assay Details
| Gene Symbol: IFI27L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076061.2 | NP_001069529.1 | 1 - 2 | 38 | 116 | |
| GenBank mRNA | BC114186.1 | - | 1 - 2 | 40 | 116 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC114186.1, NM_001076061.2 transcripts.
Set Membership: |
|

IFI27L2
Genomic Map