Assay Details
| Gene Symbol: AP2S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076078.2 | NP_001069546.1 | 3 - 4 | 330 | 87 | |
| GenBank mRNA | BC118437.1 | - | 3 - 4 | 331 | 87 | |
More Information
Set Membership: |
|

AP2S1
Genomic Map