Assay Details
| Gene Symbol: MACROH2A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076086.1 | NP_001069554.1 | 2 - 3 | 349 | 67 | |
| GenBank mRNA | BC116136.1 | - | 2 - 3 | 349 | 67 | |
More Information
Set Membership: |
|

MACROH2A2
Genomic Map