Assay Details
| Gene Symbol: TMEFF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076101.2 | NP_001069569.1 | 9 - 10 | 1441 | 69 | |
| GenBank mRNA | BC118420.1 | - | 9 - 10 | 1442 | 69 | |
More Information
Set Membership: |
|

TMEFF2
Genomic Map