Assay Details
| Gene Symbol: P4HTM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076115.2 | NP_001069583.1 | 6 - 7 | 1062 | 75 | |
| GenBank mRNA | BC114843.1 | - | 6 - 7 | 1063 | 75 | |
More Information
Set Membership: |
|

P4HTM
Genomic Map