Assay Details
| Gene Symbol: CHST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076145.1 | NP_001069613.1 | 1 - 2 | 168 | 73 | |
| GenBank mRNA | BC116070.1 | - | 1 - 2 | 168 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC116070.1, NM_001076145.1 transcripts.
Set Membership: |
|

CHST4
Genomic Map