Assay Details
| Gene Symbol: CNRIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076183.1 | NP_001069651.1 | 1 - 2 | 551 | 72 | |
| GenBank mRNA | BC118266.1 | - | 1 - 2 | 551 | 72 | |
More Information
Set Membership: |
|

CNRIP1
Genomic Map