Assay Details
| Gene Symbol: LANCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076227.1 | NP_001069695.1 | 5 - 6 | 966 | 63 | |
| GenBank mRNA | BC115991.1 | - | 5 - 6 | 966 | 63 | |
More Information
Set Membership: |
|

LANCL1
Genomic Map