Assay Details
| Gene Symbol: TRPM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076236.2 | NP_001069704.2 | 4 - 5 | 805 | 66 | |
| GenBank mRNA | BC118157.1 | - | 4 - 5 | 576 | 66 | |
More Information
Set Membership: |
|

TRPM3
Genomic Map