Assay Details
| Gene Symbol: GRWD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076239.1 | NP_001069707.1 | 6 - 7 | 1027 | 78 | |
| GenBank mRNA | BC116037.1 | - | 6 - 7 | 1027 | 78 | |
More Information
Set Membership: |
|

GRWD1
Genomic Map