Assay Details
| Gene Symbol: SPSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076280.1 | NP_001069748.1 | 1 - 2 | 147 | 95 | |
| GenBank mRNA | BC118107.1 | - | 1 - 2 | 147 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118107.1, NM_001076280.1 transcripts.
Set Membership: |
|

SPSB2
Genomic Map