Assay Details
| Gene Symbol: ARPIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076304.1 | NP_001069772.1 | 1 - 2 | 151 | 116 | |
| GenBank mRNA | BC118178.1 | - | 1 - 2 | 151 | 116 | |
More Information
Set Membership: |
|

ARPIN
Genomic Map