Assay Details
| Gene Symbol: B3GNT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076342.1 | NP_001069810.1 | 1 - 2 | 90 | 107 | |
| GenBank mRNA | BC118097.1 | - | 1 - 2 | 90 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118097.1, NM_001076342.1 transcripts.
Set Membership: |
|

B3GNT9
Genomic Map