Assay Details
| Gene Symbol: MAD2L1BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076381.2 | NP_001069849.1 | 1 - 2 | 75 | 71 | |
| GenBank mRNA | BC118446.1 | - | 1 - 2 | 76 | 71 | |
More Information
Set Membership: |
|

MAD2L1BP
Genomic Map