Assay Details
| Gene Symbol: HSPB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076412.1 | NP_001069880.1 | 5 - 6 | 434 | 95 | |
| GenBank mRNA | BC118487.1 | - | 5 - 6 | 434 | 95 | |
More Information
Set Membership: |
|

HSPB11
Genomic Map