Assay Details
| Gene Symbol: SPIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076421.1 | NP_001069889.1 | 4 - 5 | 301 | 98 | |
| GenBank mRNA | BC114834.1 | - | 4 - 5 | 301 | 98 | |
More Information
Set Membership: |
|

SPIC
Genomic Map