Assay Details
| Gene Symbol: PRXL2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076436.2 | NP_001069904.1 | 1 - 2 | 237 | 71 | |
| GenBank mRNA | BC118425.1 | - | 1 - 2 | 239 | 71 | |
More Information
Set Membership: |
|

PRXL2C
Genomic Map