Assay Details
| Gene Symbol: CCSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076437.1 | NP_001069905.1 | 2 - 3 | 1610 | 95 | |
| GenBank mRNA | BC114744.1 | - | 2 - 3 | 1610 | 95 | |
More Information
Set Membership: |
|

CCSER1
Genomic Map