Assay Details
| Gene Symbol: ANGPT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076483.2 | NP_001069951.1 | 2 - 3 | 1003 | 62 | |
| GenBank mRNA | BC114033.1 | - | 2 - 3 | 1005 | 62 | |
| EU599227.1 | - | 2 - 3 | 743 | 62 | ||
More Information
Set Membership: |
|

ANGPT4
Genomic Map