Assay Details
| Gene Symbol: DOC2G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076499.2 | NP_001069967.1 | 10 - 11 | 1189 | 104 | |
| GenBank mRNA | BC118129.1 | - | 10 - 11 | 1190 | 104 | |
More Information
Set Membership: |
|

DOC2G
Genomic Map