Assay Details
| Gene Symbol: DOC2G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076499.2 | NP_001069967.1 | 1 - 2 | 47 | 83 | |
| GenBank mRNA | BC118129.1 | - | 1 - 2 | 48 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118129.1, NM_001076499.2 transcripts.
Set Membership: |
|

DOC2G
Genomic Map