Assay Details
| Gene Symbol: GBGT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076509.2 | NP_001069977.1 | 6 - 7 | 534 | 62 | |
| GenBank mRNA | BC118408.1 | - | 6 - 7 | 535 | 62 | |
More Information
Set Membership: |
|

GBGT1
Genomic Map