Assay Details
| Gene Symbol: BCL2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076533.1 | NP_001070001.1 | 2 - 3 | 146 | 87 | |
| GenBank mRNA | BC114652.1 | - | 2 - 3 | 146 | 87 | |
| BT029856.1 | - | 460 | 87 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC114652.1, NM_001076533.1 transcripts.
Set Membership: |
|

BCL2L2
Genomic Map