Assay Details
| Gene Symbol: MS4A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076541.1 | NP_001070009.1 | 6 - 7 | 2916 | 152 | |
| GenBank mRNA | BC114037.1 | - | 6 - 7 | 2916 | 152 | |
More Information
Set Membership: |
|

MS4A14
Genomic Map