Assay Details
| Gene Symbol: HASPIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076544.2 | NP_001070012.1 | 1 | 2849 | 133 | |
| GenBank mRNA | BC112565.1 | - | 1 | 2850 | 133 | |
More Information
Set Membership: |
|

HASPIN
Genomic Map