Assay Details
| Gene Symbol: MRPS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077101.2 | NP_001070569.1 | 2 - 3 | 117 | 114 | |
| GenBank mRNA | BC114019.1 | - | 2 - 3 | 118 | 114 | |
More Information
Set Membership: |
|

MRPS12
Genomic Map