Assay Details
| Gene Symbol: HAVCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077105.2 | NP_001070573.1 | 2 - 3 | 460 | 109 | |
| GenBank mRNA | AB689695.1 | - | 2 - 3 | 427 | 109 | |
| BC114846.1 | - | 2 - 3 | 461 | 109 | ||
| LC271182.1 | - | 2 - 3 | 399 | 109 | ||
More Information
Set Membership: |
|

HAVCR2
Genomic Map