Assay Details
| Gene Symbol: HAVCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077105.2 | NP_001070573.1 | 4 - 5 | 581 | 75 | |
| GenBank mRNA | BC114846.1 | - | 4 - 5 | 582 | 75 | |
| LC271182.1 | - | 4 - 5 | 520 | 75 | ||
More Information
Set Membership: |
|

HAVCR2
Genomic Map