Assay Details
| Gene Symbol: HIGD1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077114.1 | NP_001070582.1 | 1 | 36 | 135 | |
| GenBank mRNA | BC116168.1 | - | 1 | 36 | 135 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC116168.1, NM_001077114.1 transcripts.
Set Membership: |
|

HIGD1A
Genomic Map