Assay Details
| Gene Symbol: CIBAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077134.2 | NP_001070602.1 | 1 - 2 | 150 | 72 | |
| GenBank mRNA | BC114851.1 | - | 1 - 2 | 151 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC114851.1, NM_001077134.2 transcripts.
Set Membership: |
|

CIBAR2
Genomic Map