Assay Details
| Gene Symbol: CIBAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077134.2 | NP_001070602.1 | 6 - 7 | 660 | 91 | |
| GenBank mRNA | BC114851.1 | - | 6 - 7 | 661 | 91 | |
More Information
Set Membership: |
|

CIBAR2
Genomic Map