Assay Details
| Gene Symbol: BCL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078028.1 | NP_001071496.1 | 1 - 2 | 279 | 159 | |
| GenBank mRNA | BC118326.1 | - | 1 - 2 | 279 | 159 | |
More Information
Set Membership: |
|

BCL10
Genomic Map