Assay Details
| Gene Symbol: PIGB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078055.1 | NP_001071523.1 | 11 - 12 | 1527 | 85 | |
| GenBank mRNA | BC116127.1 | - | 11 - 12 | 1527 | 85 | |
More Information
Set Membership: |
|

PIGB
Genomic Map