Assay Details
| Gene Symbol: TM2D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079593.2 | NP_001073061.1 | 3 - 4 | 368 | 70 | |
| GenBank mRNA | BC112745.1 | - | 3 - 4 | 369 | 70 | |
More Information
Set Membership: |
|

TM2D1
Genomic Map