Assay Details
| Gene Symbol: TIPRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079615.2 | NP_001073083.1 | 7 - 8 | 911 | 123 | |
| GenBank mRNA | BC114024.1 | - | 7 - 8 | 914 | 123 | |
More Information
Set Membership: |
|

TIPRL
Genomic Map