Assay Details
| Gene Symbol: TIPRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079615.2 | NP_001073083.1 | 8 - 9 | 975 | 136 | |
| GenBank mRNA | BC114024.1 | - | 8 - 9 | 978 | 136 | |
More Information
Set Membership: |
|

TIPRL
Genomic Map