Assay Details
| Gene Symbol: GIPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080279.1 | NP_001073748.1 | 1 - 2 | 258 | 72 | |
| GenBank mRNA | BC116034.1 | - | 1 - 2 | 258 | 72 | |
More Information
Set Membership: |
|

GIPC2
Genomic Map