Assay Details
| Gene Symbol: CCDC106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080334.1 | NP_001073803.1 | 1 - 2 | 41 | 72 | |
| GenBank mRNA | BC116140.1 | - | 1 - 2 | 41 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC116140.1, NM_001080334.1 transcripts.
Set Membership: |
|

CCDC106
Genomic Map