Assay Details
| Gene Symbol: CCDC106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080334.1 | NP_001073803.1 | 5 - 6 | 596 | 137 | |
| GenBank mRNA | BC116140.1 | - | 5 - 6 | 596 | 137 | |
More Information
Set Membership: |
|

CCDC106
Genomic Map