Assay Details
| Gene Symbol: KCTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080360.2 | NP_001073829.2 | 4 - 5 | 2696 | 75 | |
| GenBank mRNA | BC113314.1 | - | 4 - 5 | 696 | 75 | |
More Information
Set Membership: |
|

KCTD1
Genomic Map