Assay Details
| Gene Symbol: MAGEH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080728.1 | NP_001074197.1 | 1 | 1253 | 130 | |
| GenBank mRNA | BC116048.1 | - | 1 | 1253 | 130 | |
More Information
Set Membership: |
|

MAGEH1
Genomic Map