Assay Details
| Gene Symbol: RPRM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080739.1 | NP_001074208.1 | 1 | 1351 | 71 | |
| GenBank mRNA | BC114727.1 | - | 1 | 1351 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPRM| Gene Symbol: RPRM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080739.1 | NP_001074208.1 | 1 | 1351 | 71 | |
| GenBank mRNA | BC114727.1 | - | 1 | 1351 | 71 | |
Set Membership: |
|