Assay Details
| Gene Symbol: NT5E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174129.4 | NP_776554.2 | 3 - 4 | 786 | 62 | |
| GenBank mRNA | BC114093.1 | - | 3 - 4 | 787 | 62 | |
| BT026240.1 | - | 3 - 4 | 783 | 62 | ||
| D14541.1 | - | 3 - 4 | 877 | 62 | ||
| S64302.1 | - | 3 - 4 | 877 | 62 | ||
More Information
Set Membership: |
|

NT5E
Genomic Map