Assay Details
| Gene Symbol: ROM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174174.3 | NP_776599.2 | 1 - 2 | 790 | 68 | |
| GenBank mRNA | BC118152.1 | - | 1 - 2 | 790 | 68 | |
| D83385.1 | - | 1 - 2 | 600 | 68 | ||
| U72027.1 | - | 1 - 2 | 585 | 68 | ||
More Information
Set Membership: |
|

ROM1
Genomic Map